- Biomimetic synthesis of xuxuarines Ealpha and Ebeta: structure revision of Rzedowskia bistriterpenoids.
Biomimetic synthesis of xuxuarines Ealpha and Ebeta: structure revision of Rzedowskia bistriterpenoids.
Bioorganic & medicinal chemistry (2007-12-07)
Neil E Jacobsen, E M Kithsiri Wijeratne, Joaquim Corsino, Maysa Furlan, Vanderlan da S Bolzani, A A Leslie Gunatilaka
PMID18055208
ABSTRACT
Reaction of pristimerin with 2,3-dichloro-5,6-dicyano-1,4-benzoquinone (DDQ) resulted in a biomimetic-type coupling leading to xuxuarines Ealpha and Ebeta and not the previously reported Rzedowskia bistriterpenoids I and II suggesting that the structures proposed for these natural products need revision. A product obtained in this reaction by an unusual Diels-Alder addition followed by retro-Diels-Alder-type elimination was characterized as pristimerin dicyanophenalenedione. Complete 1H, and 13C NMR spectral assignments of xuxuarines Ealpha and Ebeta have been made by the application of 1D and 2D NMR techniques.