- Triple channel sensing of Pb(II) ions by a simple multiresponsive ferrocene receptor having a 1-deazapurine backbone.
Triple channel sensing of Pb(II) ions by a simple multiresponsive ferrocene receptor having a 1-deazapurine backbone.
Organic letters (2007-12-07)
Fabiola Zapata, Antonio Caballero, Arturo Espinosa, Alberto Tarraga, Pedro Molina
PMID18052288
ABSTRACT
A new redox, chromogenic, and fluorescent chemosensor molecule based on a deazapurine ring selectively senses aqueous Pb2+ in acetonitrile over other metal ions examined: redox shift (DeltaE1/2 = 0.15 V of the Fe(II)/Fe(III) redox couple), the colorless to orange color change, and an emission change of 620-fold, with an unprecedented detection limit of 2.7 microg L-1. The signal transduction occurs via a reversible CHEF with this inherent quenching metal ion.