- HPLC-MSn and GC-MS methods to study sunlight and UV-lamp degradations of 1-amino-5-naphthalene sulfonate.
HPLC-MSn and GC-MS methods to study sunlight and UV-lamp degradations of 1-amino-5-naphthalene sulfonate.
Chemosphere (2007-01-26)
V Gianotti, F Gosetti, S Polati, M C Gennaro
PMID17250870
ABSTRACT
HPLC-DAD, HPLC-MS/MS, GC-MS and spectrophotometric methods are employed to investigate the degradation process of sodium 1-amino-5-naphthalene sulfonate (1A5NS) aqueous solutions, when exposed to sunlight and UV-lamp (254 nm) irradiations. Experimental results show that both sunlight and 254 nm UV-lamp irradiations destroy the chemical and give rise to major degradation products, characterised by the same m/z ratios. Degradation times are lower for sunlight irradiation, for which a t(1/2) value of 137.4 min has been evaluated, in comparison with the value of t(1/2) of 26.8 min, observed for UV-lamp irradiation. The degradation pathway and the structures of the degradation products are proposed.