Merck
  • Activity of 9-nitro-camptothecin, an oral topoisomerase I inhibitor, in myelodysplastic syndrome and chronic myelomonocytic leukemia.
Activity of 9-nitro-camptothecin, an oral topoisomerase I inhibitor, in myelodysplastic syndrome and chronic myelomonocytic leukemia.
Cancer (2006-09-07)
Alfonso Quintas-Cardama, Hagop Kantarjian, Susan O'Brien, Elias Jabbour, Francis Giles, Farhad Ravandi, Stefan Faderl, Sherry Pierce, Jenny Shan, Srdan Verstovsek, Jorge Cortes
ABSTRACT
Topoisomerase I inhibitors, like topotecan, have activity in myelodysplastic syndrome (MDS) and chronic myelomonocytic leukemia (CMML). 9-Nitro-camptothecin (9-NC) is a new oral topoisomerase inhibitor with a good safety profile. The aims of the current study were to evaluate the activity and safety of 9-NC in MDS and CMML. Adults with a diagnosis of MDS (n = 12) and CMML (n = 32) received 9-NC 2 mg/m(2) orally daily 5 days a week, every 4 to 6 weeks. Overall, 5 (11%) patients achieved complete response (CR), 7 (16%) had a partial response (PR), and 6 (14%) had hematologic improvement (HI), for an overall response rate of 41%. The response rate was similar in MDS and CMML. Severe (Grade 3-4) side effects included nausea and vomiting (7%), diarrhea (18%), other gastrointestinal toxicities (5%), and genitourinary toxicities (12%). 9-NC is active in MDS and CMML. The paucity of available therapies in CMML makes 9-NC a good candidate for further studies as a single agent, or in combination with decitabine, 5-azacitidine or cytarabine.