- Studies on the interaction between Oxaprozin-E and bovine serum albumin by spectroscopic methods.
Studies on the interaction between Oxaprozin-E and bovine serum albumin by spectroscopic methods.
International journal of biological macromolecules (2006-07-11)
Shao-Fa Sun, Bo Zhou, Han-Na Hou, Yi Liu, Guang-Ya Xiang
PMID16828154
ABSTRACT
The interaction between Oxaprozin-E and bovine serum albumin (BSA) was studied by spectroscopic methods including fluorescence and UV-vis absorption spectroscopy. The quenching mechanism of fluorescence of BSA by Oxaprozin-E was discussed to be a dynamic quenching procedure. The number of binding sites n and apparent binding constant K was measured by fluorescence quenching method. The thermodynamics parameter DeltaH, DeltaG, DeltaS were calculated. The results indicate the binding reaction was mainly entropy-driven and hydrophobic forces played major role in the binding reaction. The distance r between donor (BSA) and acceptor (Oxaprozin-E) was obtained according to Förster theory of non-radioactive energy transfer.