Merck
  • Alpha1D-adrenoceptor-induced relaxation on rat carotid artery is impaired during the endothelial dysfunction evoked in the early stages of hyperhomocysteinemia.
Alpha1D-adrenoceptor-induced relaxation on rat carotid artery is impaired during the endothelial dysfunction evoked in the early stages of hyperhomocysteinemia.
European journal of pharmacology (2006-07-11)
Claudia Roberta de Andrade, Sandra Yasuyo Fukada, Vania Claudia Olivon, Marcio A F de Godoy, Renato Haddad, Marcos Nogueira Eberlin, Fernando Queiroz Cunha, Heraldo Possolo de Souza, Francisco R M Laurindo, Ana Maria de Oliveira
ABSTRACT
Hyperhomocysteinemia is a known risk factor for cardiovascular diseases, but the underlying mechanisms of this pathology are complex. We aimed to evaluate the effect of hyperhomocysteinemia in vasorelaxations induced by alpha(1D)-adrenoceptor agonists. Vascular reactivity of rat carotid artery to the alpha-adrenoceptor agonist, phenylephrine, was enhanced in hyperhomocysteinemia. Mechanical removal of endothelium did not modify the carotid responsiveness to phenylephrine, compared to control. Phenylephrine induces endothelium-dependent relaxation, in the presence of 5-methyl urapidil (alpha(1A)-adrenoceptor antagonist). We hypothesised that endothelial-relaxant alpha(1)-adrenoceptors are impaired by hyperhomocysteinemia. Incubation with prazosin (selective alpha(1)-adrenoceptor antagonist) or BMY7378 (8-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-8-azaspiro[4,5]decane-7, 9-dione dihydrochloride) (selective alpha(1D)-adrenoceptor antagonist), similarly inhibited phenylephrine-induced relaxations in both control and hyperhomocysteinemic carotids. Immunohistochemistry showed enhanced immunoreactivity for eNOS and iNOS in hyperhomocysteinemic rats. In carotid arteries from hyperhomocysteinemic rats there was a decrease in superoxide dismutase activity and enhanced superoxide anion production. We conclude that alpha(1D)-adrenoceptors mediate endothelium-dependent relaxation triggered by phenylephrine in rat carotid artery and affect the final tone. Furthermore, the enhanced phenylephrine-induced contraction in carotid artery due to hyperhomocysteinemia is endothelium-dependent and involves a loss of the inhibitory effect of relaxant alpha(1D)-adrenoceptors by reducing NO biodisponibility.