Merck
  • Differential effects of selective and non-selective inhibition of nitric oxide synthase on the expression and activity of cyclooxygenase-2 during gastric ulcer healing.
Differential effects of selective and non-selective inhibition of nitric oxide synthase on the expression and activity of cyclooxygenase-2 during gastric ulcer healing.
European journal of pharmacology (2006-04-08)
Jin Sheng Guo, Chi Hin Cho, Ji Yao Wang, Marcel Wing Leung Koo
ABSTRACT
Nitric oxide synthases (NOS) and cyclooxygenase-2 (COX-2) are important enzymes involved in ulcer healing but interactions between them have not been clearly defined. The aim of this study was to investigate the effects of selective or non-selective inhibition of NOS on the expression and activity of COX-2 during healing of acetic acid-induced gastric ulcers in rats. N-[3-(aminomethyl)benzyl] acetamidine (1400 W), a potent selective inhibitor of inducible nitric oxide synthase (iNOS), at a dose of 0.1 mg/kg/day, was found to reduce the ulcer sizes at day 3 and 7 post-ulcer induction. On the other hand, 15 mg/kg/day of NG-nitro-L-arginine methyl ester (L-NAME), a non-selective NOS inhibitor that suppresses both iNOS and endothelial nitric oxide synthase (eNOS), enlarged the ulcer sizes over the same time periods. The expression of COX-2 and COX activity, together with NF-kappaB activation in the ulcer tissues were down-regulated by L-NAME but not 1400 W. It is concluded that iNOS may contribute to ulcer formation while COX-2 and eNOS promote ulcer healing. eNOS enhances COX-2 expression possibly through the activation of NF-kappaB.