Merck
  • Diels-Alder reactions of cyclopentadiene and 9,10-dimethylanthracene with cyanoalkenes: the performance of density functional theory and Hartree-Fock calculations for the prediction of substituent effects.
Diels-Alder reactions of cyclopentadiene and 9,10-dimethylanthracene with cyanoalkenes: the performance of density functional theory and Hartree-Fock calculations for the prediction of substituent effects.
The journal of physical chemistry. A (2006-01-27)
Gavin O Jones, Vildan A Guner, K N Houk
ABSTRACT
The activation enthalpies for the reactions of cyclopentadiene with ethylene, acrylonitrile, fumaronitrile, maleonitrile, vinylidene cyanide, tricyanoethylene, and tetracyanoethylene have been computed with the Hartree-Fock quantum mechanical method and a variety of density functionals (B3LYP, BPW91, and MPW1K) and compared to classic experimental data obtained by Sauer, Wiest, and Mielert (Chem. Ber. 1964, 97, 3183). Some significant errors in B3LYP activation enthalpies involving cyano groups, especially 3 or 4 cyano groups, have been identified, whereas HF and MPW1K reproduce substituent effects quite accurately.