Merck
  • Aerial optimization and canopy penetration study of Dibrom 14 Concentrate.
Aerial optimization and canopy penetration study of Dibrom 14 Concentrate.
Journal of the American Mosquito Control Association (2005-04-14)
James R Brown, W H Reynolds, C Palmisano, V Taylor
ABSTRACT
This document describes the results of collaborative trials between St. Tammany Parish Mosquito Control, ADAPCO, Inc., and the Navy Disease Vector Ecology and Control Center, Naval Air Station, Jacksonville, FL. These tests provide data on the aerial optimization testing of Dibrom 14 Concentrate adulticide. During the week of June 17, 2002, 1 canopy penetration and 3 optimization studies were conducted in St. Tammany Parish, LA, using a Britton Norman twin turbine Islander equipped with 10-800067 flat fan nozzles. Dibrom 14 Concentrate (AMVAC Chemical Corp., Los Angeles, CA) was applied at 3.12 liter per min from a release height of 60.96 m above ground level (agl) at 140 kts indicated airspeed. This resulted in an application rate of 0.65 oz/acre.