Merck
  • [Studies on chemical constituents in herb of Lamium maculatum var. kansuense (II)].
[Studies on chemical constituents in herb of Lamium maculatum var. kansuense (II)].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2005-02-24)
Yan-ru Deng, Lan Ding, Shui-xian Wu, Han-qing Wang
ABSTRACT
To study the chemical constituents from Lamium maculatum var. kansuense. The chemical constituents were isolated and repeatedly purified on silica gel column and the structures were elucidated by the NMR spectra and physico-chemical properties. Six compounds were obtained and identified as polypodine B (I), 5-OH-8-epiloganin (II), shlanzhiside methyl ester (III), liriodendrin (IV), quercitroside (V), uridine (VI). Compound IV was found from genus Lamium for the first time and the rest of the compounds were found from Lamium maculatum var kansuense for the first time.