Merck
  • Three thiamine analogues differently alter thiamine transport and metabolism in nervous tissue: an in vivo kinetic study using rats.
Three thiamine analogues differently alter thiamine transport and metabolism in nervous tissue: an in vivo kinetic study using rats.
Metabolic brain disease (2004-05-07)
G Rindi, C Patrini, A Nauti, R Bellazzi, P Magni
ABSTRACT
Thiamine (T) analogues pyrithiamine, oxythiamine or amprolium in amounts 10-1000 times higher than labelled T, were i.p. injected into rats together with 14C-T (30 microg; 46.25 KBq). The radioactivity associated with T and its phosphoesters in the plasma and cerebral cortex, brainstem, cerebellum, and sciatic nerve were determined at time intervals from 0.25 to 240 h from injection. The experimental data obtained were processed with a mathematical compartmental model that calculated the fractional rate constants. These are the amount of content in a given compartment that is replaced in 1 h and expressed in per hour. The results showed that all three analogues inhibited thiamine entry from plasma. Instead, oxythiamine enhanced T phosphorylation to T pyrophosphate (TPP); amprolium and oxythiamine enhanced TPP dephosphorylation to monophosphate (TMP); pyrithiamine reduced TPP dephosphorylation and TMP formation, while none of the analogues modified TMP dephosphorylation to T. In conclusion, in living rats, the action of T analogues was much more complex than could be expected from their structure and action in vitro.