Merck
  • Minimizing the impact of the mosquito adulticide naled on honey bees, Apis mellifera (Hymenoptera: Apidae): aerial ultra-low-volume application using a high-pressure nozzle system.
Minimizing the impact of the mosquito adulticide naled on honey bees, Apis mellifera (Hymenoptera: Apidae): aerial ultra-low-volume application using a high-pressure nozzle system.
Journal of economic entomology (2004-03-05)
He Zhong, Mark Latham, Steve Payne, Cate Brock
ABSTRACT
The impact of the mosquito adulticide naled on honey bees, Apis mellifera L., was evaluated by exposing test beehives to nighttime aerial ultra-low-volume (ULV) applications using a high-pressure nozzle system. The tests were conducted during routine mosquito control missions at Manatee County, Florida, in summer 2000. Two treatment sites were sprayed a total of four times over a 10-wk period. Honey bees, which clustered outside of the hive entrances, were subjected to naled exposure during these mosquito control sprays. The highest average naled ground deposition was 2,688 microg/m2 at the Port Manatee site, which resulted in statistically significant bee mortality (118) compared with the controls. At the Terra Ceia Road site, an intermediate level of naled deposition was found (1,435 microg/m2). For this spray mission, the range of dead bees per hive at Terra Ceia was 2 to 9 before spraying and 5 to 36 after naled application. Means of all other naled ground depositions were < 850 microl/m2. We concluded that substantial bee mortality (> 100 dead bees) resulted when naled residue levels were > 2,000 kg/m2 and honey bees were clustered outside of the hive entrances during mosquito adulticide applications. Compared with the flat-fan nozzle systems currently used by most of Florida's mosquito control programs, the high-pressure nozzle system used in this experiment substantially reduced environmental insecticide contamination and lead to decreased bee mortality. Statistical analysis also showed that average honey yield at the end of the season was not significantly reduced for those hives that were exposed to the insecticide.