Merck
  • Mutagenicity of substituted anthraquinones in the Ames/Salmonella microsome system.
Mutagenicity of substituted anthraquinones in the Ames/Salmonella microsome system.
Mutation research (1992-05-01)
S Krivobok, F Seigle-Murandi, R Steiman, D R Marzin, V Betina
ABSTRACT
Unsubstituted anthraquinone, 4 substituted anthraquinones (emodin, danthron, physcion, a new compound M-108-C) and 3 dimers (skyrin, rugulosin, rugulin) were tested using the Ames/Salmonella assay (strains TA98, TA100, TA1537 and TA102). Danthron and emodin were found to be mutagenic for TA1537 with or without metabolic activation, physcion only with metabolic activation. A significant difference was found between the mutagenic activities of emodin (16.2 His+/nmole) and danthron (6.5 His+/nmole) as well as a high specific mutagenic activity for physcion (11.6 His+/nmole). These results on structure-mutagenic activity relationships suggest that the 6-methyl group plays an important role in the mutagenic activity after metabolic activation. Furthermore, and contrary to emodin, physcion exhibited a weak mutagenic activity for TA102, probably due to the formation of a different metabolite. Such information is necessary to evaluate the potential carcinogenic hazard of these compounds.