- Anorexigen-related cardiopulmonary toxicity.
Anorexigen-related cardiopulmonary toxicity.
Reviews in cardiovascular medicine (2002-11-29)
S B Gross, N E Lepor
PMID12457145
ABSTRACT
Three years after the withdrawal of fenfluramine and dexfenfluramine from the market, the magnitude and prevalence of their deleterious cardiopulmonary effects remain undetermined. The links between these anorexigens and valvular heart disease and primary pulmonary hypertension, however, are clearly established. Because some evidence indicates that the valvular lesions may regress with cessation of the drug, management guidelines are still in flux. Patient reassurance and close surveillance, including serial echocardiography in selected cases, are warranted.