Merck
  • Distinction between esterases and lipases: a kinetic study with vinyl esters and TAG.
Distinction between esterases and lipases: a kinetic study with vinyl esters and TAG.
Lipids (2002-09-10)
Henri Chahinian, Lylia Nini, Elisabeth Boitard, Jean-Paul Dubès, Louis-Claude Comeau, Louis Sarda
ABSTRACT
The better to characterize enzymes hydrolyzing carboxyl ester bonds (carboxyl ester hydrolases), we have compared the kinetic behavior of various lipases and esterases against solutions and emulsions of vinyl esters and TAG. Short-chain vinyl esters are hydrolyzed at comparable rates by esterases and lipases and have higher limits of solubility in water than corresponding TAG. Therefore, they are suited to study the influence of the physical state of the substrate on carboxyl ester hydrolase activity within a large concentration range. Enzymes used in this study are TAG lipases from microorganisms, lipases from human and guinea pig pancreas, pig liver esterase, and acetylcholinesterase. This study also includes cutinase, a fungal enzyme that displays functional properties between esterases and lipases. Esterases display maximal activity against solutions of short-chain vinyl esters (vinyl acetate, vinyl propionate, and vinyl butyrate) and TAG (triacetin, tripropionin, and tributyrin). Half-maximal activity is reached at ester concentrations far below the solubility limit. The transition from solution to emulsion at substrate concentrations exceeding the solubility limit has no effect on esterase activity. Lipases are active on solutions of short-chain vinyl esters and TAG but, in contrast to esterases, they all display maximal activity against emulsified substrates and half-maximal activity is reached at substrate concentrations near the solubility limit of the esters. The kinetics of hydrolysis of soluble substrates by lipases are either hyperbolic or deviate from the Michaelis-Menten model and show no or weak interfacial activation. The presence of molecular aggregates in solutions of short-chain substrates, as evidenced by a spectral dye method, likely accounts for the activity of lipases against soluble esters. Unlike esterases, lipases hydrolyze emulsions of water-insoluble medium- and long-chain vinyl esters and TAG such as vinyl laurate, trioctanoin, and olive oil. In conclusion, comparisons of the kinetic behavior of carboxyl ester hydrolases against solutions and emulsions of vinyl esters and TAG allows the distinction between lipases and esterases. In this respect, it clearly appears that guinea pig pancreatic lipase and cutinase are unambiguously classified as lipases.
Sigma-Aldrich
Tripropionin, ≥97.1%, FCC, FG