Merck
  • Biphasic effects of oxethazaine, a topical anesthetic, on the intracellular Ca(2+) concentration of PC12 cells.
Biphasic effects of oxethazaine, a topical anesthetic, on the intracellular Ca(2+) concentration of PC12 cells.
Biochemical pharmacology (2002-08-09)
Yasusuke Masuda, Tamami Oguma, Akira Kimura
ABSTRACT
There have been few reports on the mechanism(s) of action of oxethazaine (OXZ) despite its potent local anesthetic action. Generally, local anesthetics (LAs) not only inhibit Na(+) channels but also affect various membrane functions. In the present study, using PC12 cells as a nerve cell model, the effects of OXZ on intracellular Ca(2+) concentration ([Ca(2+)](i)) were examined in relation to cytotoxicity and dopamine release. [Ca(2+)](i) was determined by the quin2 method. In resting cells, (6-10)x10(-5)M OXZ produced lactate dehydrogenase leakage, which was Ca(2+)-dependent, inhibited by metal Ca(2+) channel blockers, and preceded by a marked increase in [Ca(2+)](i). Some other LAs showed no cytotoxicity at these concentrations. In K(+)-depolarized cells, however, lower concentrations of OXZ (10(-6)-10(-7)M), that had no effect on resting [Ca(2+)](i), inhibited both the dopamine release and the increase of [Ca(2+)](i) in parallel. The inhibitory potency against the [Ca(2+)](i) increase was in the order of nifedipine>OXZ approximately verapamil>diltiazem, and OXZ acted additively on the Ca(2+) channel blockers. OXZ showed the least effect on K(+)-depolarization as determined by bisoxonol uptake. OXZ also inhibited the increase in [Ca(2+)](i) induced by S(-)-BAY K 8644, a Ca(2+) channel agonist. These observations suggested that low concentrations of OXZ interact with L-type Ca(2+) channels. The biphasic effects of OXZ on Ca(2+) movement may be due to a unique chemical structure, and may participate in and complicate the understanding of the potent pharmacological and toxicological actions of OXZ.