Merck
  • Determination of oxanilic and sulfonic acid metabolites of acetochlor in soils by liquid chromatography-electrospray ionisation mass spectrometry.
Determination of oxanilic and sulfonic acid metabolites of acetochlor in soils by liquid chromatography-electrospray ionisation mass spectrometry.
Journal of chromatography. A (2002-07-10)
T Dagnac, R Jeannot, C Mouvet, N Baran
ABSTRACT
An analytical method is presented that describes the extraction and quantification of oxanilic and sulfonic acid metabolites of the herbicide acetochlor in soil samples. Experiments were performed on 50 g of soil using a solvent extraction technique with an acetonitrile-water (60:40) mixture in an acidic medium. Analysis was carried out by reversed-phase liquid chromatography and detection by electrospray ionisation mass spectrometry in single ion monitoring and negative modes. Four different soil matrices were spiked in triplicate with standards of each degradation compound at three concentration levels between 2 and 80 microg/kg. The average recoveries range from 90 to 120% for both the metabolites, with relative standard deviations lower than 15%. The limits of quantification are about 1 and 2 microg/kg for the ethanesulfonic acid and the oxanilic acid metabolites, respectively. The method has been applied to soils and solids recovered from the deeper unsaturated zone of a small French catchment closely monitored as part of the European project "Pesticides in European Groundwaters: detailed study of Aquifers and Simulation of possible Evolution scenarios (PEGASE)".