Merck
  • Enantioselectivity of debrisoquine 4-hydroxylation in Brazilian Caucasian hypertensive patients phenotyped as extensive metabolizers.
Enantioselectivity of debrisoquine 4-hydroxylation in Brazilian Caucasian hypertensive patients phenotyped as extensive metabolizers.
Journal of chromatography. B, Biomedical sciences and applications (2001-01-06)
P M Cerqueira, F H Mateus, E J Cesarino, P S Bonato, V L Lanchote
ABSTRACT
Debrisoquine (D), an antihypertensive drug metabolized to 4-hydroxydebrisoquine (4-OHD) by CYP2D6, is commonly used as an in vivo probe of CYP2D6 activity and can be used to phenotype individuals as either extensive (EMs) or poor metabolizers (PMs) of such drugs as beta-adrenergic blockers, tricyclic antidepressants, and class 1C antiarrhythmics. This report describes reversed-phase HPLC systems by which D and 4-OHD or S-(+) and R-(-)-4-OHD in urine are more selectively quantified without the need for derivatization techniques. We also studied the urinary excretion of R-(-)- and S-(+)-4-hydroxydebrisoquine in EM hypertensive patients in order to determine weather 4-OHD formation exhibits enantioselectivity. Twelve patients with mild to severe essential hypertension were admitted to the study. They received a single tablet of Declinax containing 10 mg debrisoquine sulfate. All the urine excreted during the following 8 h was collected. The debrisoquine metabolic ratio (DMR) was calculated as % of dose excreted as D/% of dose excreted as 4-OHD and the debrisoquine recovery ratio (DRR) was calculated as % of dose excreted as 4-OHD/% of dose excreted as D+4-OHD. Debrisoquine and its metabolite were determined in urine by HPLC using a reversed-phase Select B LiChrospher column, a mobile phase of 0.25 N acetate buffer, pH 5-acetonitrile (9:1, v/v) and a fluorescence detector. The limit of quantitation was determined to be 25.0 ng/ml for D and 18.75 ng/ml for 4-OHD. Intra- and inter-day relative standard deviations (RSDs) were less than 10%. All hypertensive patients studied showed a DMR of less than 12.6 or a DRR higher than 0.12 and were classified as EMs. Direct enantioselective separation on chiral stationary phase involved resolution of S-(+)-4-OHD and R-(-)-4-OHD on a Chiralcel OD-R column with a mobile phase of 0.125 N sodium perchlorate, pH 5-acetonitrile-methanol (85:12:3, v/v/v). The quantitation limit of each enantiomer was 3.75 ng/ml of urine. Intra- and inter-day RSDs were less than 10% for each enantiomer. A high degree of enantioselectivity in the 4-hydroxylation of D favouring the S-(+) enantiomer was observed, resulting in R-(-)-4-OHD not detected in the urine of the EM hypertensive patients studied.