Merck
  • In vivo production of nitric oxide after administration of cyclohexanone oxime.
In vivo production of nitric oxide after administration of cyclohexanone oxime.
Chemical research in toxicology (1999-10-20)
R E Glover, J T Corbett, L T Burka, R P Mason
ABSTRACT
Cyclohexanone oxime (CHOX), an intermediate used in the synthesis of polycaprolactam (Nylon-6), has been reported to be hematotoxic in Fischer rats. The in vivo metabolism of CHOX was found to release nitric oxide, which was detected in venous blood by electron paramagnetic resonance spectroscopy as the nitrosylhemoglobin complex. In vitro incubation of CHOX with venous blood resulted in the formation of the characteristic nitrosylhemoglobin complex, suggesting that the blood was a possible site for metabolism. Excessive nitric oxide production may, in part, contribute to the observed toxicity of CHOX.