Merck
  • Improvement of the immune response to foot and mouth disease virus vaccine in calves by using Avridine as adjuvant.
Improvement of the immune response to foot and mouth disease virus vaccine in calves by using Avridine as adjuvant.
Veterinary immunology and immunopathology (1999-09-18)
A M Sadir, P I Zamorano, A Romera, A Wigdorovitz, E Smitsaart, L Marangunich, C Schiappacassi, M V Borca
ABSTRACT
The epidemiological analysis of the cattle population during the eradication plan of foot and mouth disease (FMD) in Argentina clearly indicated a higher incidence of the disease in animals within their first year of age. It is important to improve the efficacy of the vaccination in those animals. In a previous report, we have shown the effect of an immunomodulator, Avridine (Avr), in the enhancement of the immune response elicited by FMD virus (FMDV) vaccines in experimental hosts [Berinstein, A., Pérez Filgueira, M., Schudel, A., Zamorano, P., Borca, M., Sadir, A.M., 1993. Avridine and LPS from Brucella ovis: effect on the memory induced by foot-and-mouth disease virus vaccination in mice. Vaccine 11, 1295-1301]. In this report, we analyze the effect of Avr in the improvement of the anti-FMDV immune response elicited in young animals immunized with a tetravalent vaccine. The anti-FMDV antibody response was evaluated using a liquid-phase blocking sandwich ELISA (LPBE) [Smitsaart, E.N., Zanelli, M., Rivera, I., Fondevila, N., Compaired, D., Maradei, E., Bianchi, T., O'Donnell, V., Schudel, A.A., 1998. Assessment using ELISA of the herd immunity levels induced in cattle by foot and mouth disease oil vaccines. Prev. Vet. Med 33, 283-296] while the cellular response was detected using an antigen specific lymphoproliferative test [Zamorano, P., Wigdorovitz, A., Chaher, M., Fernández, F., Sadir, A., Borca, M., 1994. Localization of B and T cell epitopes on a synthetic peptide containing the major immunogenic site of FMDV O1 Campos. Virology 201, 383-387]. The results show that, while no differences were detected in the cellular response, the anti-FMDV antibody reaction was significantly (<0.05) higher in animals immunized with the immunogen containing Avr. At 90 days post vaccination, 89-100% of the animals immunized with Avr presented predicted protection (PP) higher than 82% while just 50-61% of the animals immunized with vaccine without immunomodulator presented that characteristic. Also, it is shown that the increase in the anti-FMDV antibody titre in animals immunized with the vaccine containing Avr was mediated by an increase in the levels of both IgG1 and IgG2 which presented a significative correlation with LPELISA antibodies titres. It is concluded that the addition of Avr in the FMDV vaccines improve the immune status of the calves, the cattle population that suffers the highest epidemiological risk.