Merck
  • A novel role for carbonic anhydrase: cytoplasmic pH gradient dissipation in mouse small intestinal enterocytes.
A novel role for carbonic anhydrase: cytoplasmic pH gradient dissipation in mouse small intestinal enterocytes.
The Journal of physiology (1999-03-06)
A K Stewart, C A Boyd, R D Vaughan-Jones
ABSTRACT
1. The spatial and temporal distribution of intracellular H+ ions in response to activation of a proton-coupled dipeptide transporter localized at the apical pole of mouse small intestinal isolated enterocytes was investigated using intracellular carboxy-SNARF-1 fluorescence in combination with whole-cell microspectrofluorimetry or confocal microscopy. 2. In Hepes-buffered Tyrode solution, application of the dipeptide Phe-Ala (10 mM) to a single enterocyte reduced pHi locally in the apical submembranous space. After a short delay (8 s), a fall of pHi occurred more slowly at the basal pole. 3. In the presence of CO2/HCO3--buffered Tyrode solution, the apical and basal rates of acidification were not significantly different and the time delay was reduced to 1 s or less. 4. Following application of the carbonic anhydrase inhibitor acetazolamide (100 microM) in the presence of CO2/HCO3- buffer, addition of Phe-Ala once again produced a localized apical acidification that took 5 s to reach the basal pole. Basal acidification was slower than at the apical pole. 5. We conclude that acid influx due to proton-coupled dipeptide transport can lead to intracellular pH gradients and that intracellular carbonic anhydrase activity, by facilitating cytoplasmic H+ mobility, limits their magnitude and duration.