Merck
  • The ddY mouse: a model of postprandial hypertriglyceridemia in response to dietary fat.
The ddY mouse: a model of postprandial hypertriglyceridemia in response to dietary fat.
Journal of lipid research (2012-06-28)
Tomomi Yamazaki, Kyoko Kishimoto, Osamu Ezaki
ABSTRACT
Postprandial hyperlipidemia (lipemia) is a risk factor for atherosclerosis. However, mouse models of postprandial hyperlipidemia have not been reported. Here, we report that ddY mice display marked postprandial hypertriglyceridemia in response to dietary fat. In ddY mice, the fasting serum total triacylglyceride (TG) concentration was 134 mg/dl, which increased to 571 mg/dl after an intragastric safflower oil load (0.4 ml/mouse). In C57BL/6J mice, these concentrations were 57 and 106 mg/dl, respectively. By lipoprotein analysis, ddY mice showed increases in chylomicron- and VLDL-sized TG fractions (remnants and VLDL) after fat load. In C57BL/6J mice, post-heparin plasma LPL activity after fat load was increased 4.8-fold relative to fasting. However, in ddY mice, the increase of LPL activity after fat load was very small (1.2-fold) and not significant. High fat feeding for 10 weeks led to obesity in ddY mice. A difference in LPL amino acid composition between C57BL/6J and ddY mice was detected but was deemed unlikely to cause hypertriglyceridemia because hypertriglyceridemia was not evident in other strains harboring the ddY-type LPL sequence. These findings indicate that postprandial hypertriglyceridemia in ddY mice is induced by decreased LPL activity after fat load and is associated with obesity induced by a high-fat diet.