Merck
  • Liquid chromatographic fluorescence method for multiresidue determination of thiabendazole and 5-hydroxythiabendazole in milk.
Liquid chromatographic fluorescence method for multiresidue determination of thiabendazole and 5-hydroxythiabendazole in milk.
Journal of AOAC International (1995-05-01)
R V Arenas, N A Johnson
ABSTRACT
A novel liquid chromatographic multiresidue method has been developed for quantitation of thiabendazole (TBZ), the metabolite 5-hydroxythiabendazole (5-OH-TBZ), and the sulfate conjugate of 5-hydroxythiabendazole (5-HSO4-TBZ) in raw cow's milk. The 5-HSO4-TBZ is hydrolyzed quantitatively under acidic conditions to 5-OH-TBZ. TBZ and 5-OH-TBZ are extracted from milk at pH 8.0 with ethyl acetate followed by cleanup of the extract on a cation-exchange solid-phase extraction column. Analytes are quantitated by liquid chromatography with a cation-exchange stationary phase and fluorescence detection. Recoveries from raw cow's milk samples fortified with TBZ and 5-OH-TBZ or TBZ and 5-HSO4-TBZ at 0.05 to 2 ppm ranged from 87 to 103% for TBZ, 98-109% for 5-OH-TBZ, and 96-115% for 5-HSO4-TBZ (quantitated as 5-OH-TBZ). The assay provides a simple, rapid, and sensitive multiresidue method for monitoring total residues of TBZ, 5-OH-TBZ, and 5-HSO4-TBZ in milk.