- Synthesis and preliminary biological profile of new NO-donor tolbutamide analogues.
Synthesis and preliminary biological profile of new NO-donor tolbutamide analogues.
Bioorganic & medicinal chemistry letters (2012-05-09)
Yasinalli Tamboli, Loretta Lazzarato, Elisabetta Marini, Stefano Guglielmo, Michela Novelli, Pascale Beffy, Pellegrino Masiello, Roberta Fruttero, Alberto Gasco
PMID22565215
ABSTRACT
We describe a new class of NO-donor hypoglycemic products obtained by joining tolbutamide, a typical hypoglycemic sulfonylurea, with a NO-donor moiety through a hard link. As NO-donors we chose either furoxan (1,2,5-oxadiazole 2-oxide) derivatives or the classical nitrooxy function. A preliminary biological characterization of these compounds, including stimulation of insulin release from cultured rat pancreatic β-cells and in vitro vasodilator and anti-aggregatory activities, is reported.