- Supramolecular vesicle: triggered by formation of pseudorotaxane between cucurbit[6]uril and surfactant.
Supramolecular vesicle: triggered by formation of pseudorotaxane between cucurbit[6]uril and surfactant.
Chemical communications (Cambridge, England) (2011-09-20)
Qiongbo Zhou, Hao Wang, Tian Gao, Yue Yu, Bokai Ling, Lu Mao, Haibo Zhang, Xianggao Meng, Xiaohai Zhou
PMID21927725
ABSTRACT
Supramolecular vesicles are triggered by the inclusion complex formed by CB[6] and surfactant. The combination of CB[6] with the surfactant plays a key role in the vesicle formation.