Merck
  • A sensitive liquid chromatographic-mass spectrometric method for simultaneous determination of dehydroevodiamine and limonin from Evodia rutaecarpa in rat plasma.
A sensitive liquid chromatographic-mass spectrometric method for simultaneous determination of dehydroevodiamine and limonin from Evodia rutaecarpa in rat plasma.
Analytical and bioanalytical chemistry (2011-05-17)
Juanjuan Zhao, Xiao Han, Xu Zhao, Chao Wang, Qing Li, Xiaohui Chen, Kaishun Bi
ABSTRACT
A liquid chromatographic-mass spectrometric (LC-MS) method has been developed and validated for simultaneous determination of dehydroevodiamine and limonin from Evodia rutaecarpa in rat plasma. After addition of the internal standard, domperidone, plasma samples were extracted by liquid-liquid extraction with ethyl acetate and separated on an Apollo C(18) column (250 mm × 4.6 mm, 5 μm), with methanol-0.01% formic acid water (60:40, v/v) as mobile phase, within a runtime of 12.0 min. The analytes were detected without interference in the selected ion monitoring (SIM) mode with positive electrospray ionization. The linear range was 1.0-500 ng mL(-1) for dehydroevodiamine and 2.0-1,000 ng mL(-1) for limonin, with lower limits of quantitation of 1.0 and 2.0 ng mL(-1), respectively. Intra-day and inter-day precision were within 6.0% and 10.9%, respectively, for both analytes, and the accuracy (relative error, RE, %) was less than 4.8% and 6.5%, respectively. The validated method was successfully applied to a comparative pharmacokinetic study of dehydroevodiamine and limonin in rat plasma after oral administration of dehydroevodiamine, limonin, and an aqueous extract of Evodiae fructus. The results indicated there were obvious differences between the pharmacokinetic behavior after oral administration of an aqueous extract of Evodiae fructus compared with single substances.