Merck
  • Functional expression of a novel ginsenoside Rf binding protein from rat brain mRNA in Xenopus laevis oocytes.
Functional expression of a novel ginsenoside Rf binding protein from rat brain mRNA in Xenopus laevis oocytes.
Molecular pharmacology (2002-03-20)
Seok Choi, Se-Yeon Jung, Yoo-Seung Ko, Seong-Ryong Koh, Hyewhon Rhim, Seung-Yeol Nah
ABSTRACT
We have shown that ginsenoside Rf (Rf) regulates voltage-dependent Ca(2+) channels through pertussis toxin (PTX)-sensitive G proteins in rat sensory neurons. These results suggest that Rf can act through a novel G protein-linked receptor in the nervous system. In the present study, we further examined the effect of Rf on G protein-coupled inwardly rectifying K(+) (GIRK) channels after coexpression with size-fractionated rat brain mRNA and GIRK1 and GIRK4 (GIRK1/4) channel cRNAs in Xenopus laevis oocytes using two-electrode voltage-clamp techniques. We found that Rf activated GIRK channel in a dose-dependent and reversible manner after coexpression with subfractions of rat brain mRNA and GIRK1/4 channel cRNAs. This Rf-evoked current was blocked by Ba(2+), a potassium channel blocker. The size of rat brain mRNA responding to Rf was about 6 to 7 kilobases. However, Rf did not evoke GIRK current after injection with this subfraction of rat brain mRNA or GIRK1/4 channel cRNAs alone. Other ginsenosides, such as Rb(1) and Rg(1), evoked only slight induction of GIRK currents after coexpression with the subfraction of rat brain mRNA and GIRK1/4 channel cRNAs. Acetylcholine and serotonin almost did not induce GIRK currents after coexpression with the subfraction of rat brain mRNA and GIRK1/4 channel cRNAs. Rf-evoked GIRK currents were not altered by PTX pretreatment but were suppressed by intracellularly injected guanosine-5'-(2-O-thio) diphosphate, a nonhydrolyzable GDP analog. These results indicate that Rf activates GIRK channel through an unidentified G protein-coupled receptor in rat brain and that this receptor can be cloned by the expression method demonstrated here.