Merck
  • Studies of resistance to anticoccidials in Eimeria field isolates and pure Eimeria strains.
Studies of resistance to anticoccidials in Eimeria field isolates and pure Eimeria strains.
Veterinary parasitology (1997-04-01)
B Stephen, M Rommel, A Daugschies, A Haberkorn
ABSTRACT
Ten Eimeria field isolates from North Germany were studied in battery tests for sensitivity to selected anticoccidials. A high percentage of the Eimeria field isolates (9 out of 10) showed resistance to anticoccidials, mostly multiple resistance. Partial or complete resistance to maduramicin was found in 7 field isolates, to monensin in 6, to salinomycin in 5, to nicarbazin in 8, to halofuginone in 7, to robenidine and toltrazuril in 1, and to diclazuril in 2 field isolates. Multiple resistance had developed in 7 of the 10 isolates. Cross-resistance between maduramicin, monensin, and salinomycin occurred in 5 Eimeria isolates. One isolate showed cross-resistance between diclazuril and toltrazuril. From the resistant isolates 15 pure E. acerculina and 5 pure E. brunetti strains were obtained by single oocyst infections. Seven of the E. acerculina and 4 of the E. brunetti strains showed resistance or partial resistance that was also present in the original isolate. Ten of 11 resistant strains were multiply resistant.