Merck
Chromosomal mapping of core histone acetylation by immunoselection.
Methods (San Diego, Calif.) (1997-05-01)
C Crane-Robinson, T R Hebbes, A L Clayton, A W Thorne
ABSTRACT
Acetylation of specific lysine residues in the N-terminal domains of core histones is a biochemical marker of active genes. To determine the spatial and temporal distribution of this reversible posttranslational modification, affinity-purified polyclonal antibodies recognizing the epitope epsilon-acetyllysine have been used in immunoselection procedures with mononucleosomes and salt-soluble chromatin fragments generated by micrococcal nuclease. The DNA of the antibody-selected chromatin was slot-blotted and probed with a variety of gene sequences: an enhanced hybridization signal, with respect to that from the DNA of the input chromatin, demonstrated elevated acetylation levels on the histones associated with the probing sequences. Using chicken embryonic erythrocytes as chromatin source and probes from the beta globin locus, it was shown that both the embryonic epsilon and adult beta genes are acetylated at 5 and 15 days, and the acetylation uniformly covers the whole of the locus, precisely comapping with the 33 kb of open chromatin structure. Studies with proliferating human K562 cells show that the inactive but poised PDGF-beta gene is already hyperacetylated and that its acetylation status is not enhanced on induction. These results indicate that acetylation is not a consequence of transcription but a prerequisite and that it may be responsible for either generating or maintaining the open structure of poised and active genes.