- Nonionic surfactants regioselective synthesis of fatty acid esters of alpha- and beta-glucopyranose.
Nonionic surfactants regioselective synthesis of fatty acid esters of alpha- and beta-glucopyranose.
Lipids (1997-05-01)
D De Luca, A De Nino, A Liguori, A Procopio, G Sindona
PMID9168463
ABSTRACT
Lipophilic esters of saccharides belong to the family of nonionic surfactants widely employed in pharmaceutical and cosmetics formulations. A very simple method is presented whereby 6-O-esters of alpha- and beta-glucose can be prepared and isolated. Good results have been obtained in the synthesis of 6-O-oleyl derivatives by simple acylation with appropriate oleyl chloride. The condensing agent bis(2-oxo-3-oxazolidinyl) phosphinic chloride (BOP-Cl) allows the preparations of the same esters with better regioselectivity but with yields strongly dependent on the aliphatic chain length of the carboxylic acids employed.