Merck
  • Tempace and troxyl-novel synthesized 2,2,6,6-tetramethylpiperidine derivatives as antioxidants and radioprotectors.
Tempace and troxyl-novel synthesized 2,2,6,6-tetramethylpiperidine derivatives as antioxidants and radioprotectors.
Biochemistry and molecular biology international (1996-12-01)
D Metodiewa, J Skolimowski, S Karolczak
ABSTRACT
Two novel 2,2,6,6-tetramethylpiperidine derivatives (Tempace and Troxyl) were synthesized and their capacity to act as scavengers of superoxide, inhibitors of iron and ascorbate-driven Fenton reaction and radioprotectors was tested. The possibility for one-electron oxidation of novel compounds by heme-ferryl species was also examined. The results clearly indicate that Tempace and Troxyl are acting as promising antioxidants and radioprotectors thus providing a base for further investigations and pharmacological applications.