Merck
  • Evaluation of a eucalyptus-based repellent against Culicoides impunctatus (Diptera:Ceratopogonidae) in Scotland.
Evaluation of a eucalyptus-based repellent against Culicoides impunctatus (Diptera:Ceratopogonidae) in Scotland.
Journal of the American Mosquito Control Association (1996-06-01)
J K Trigg
ABSTRACT
A eucalyptus-based insect repellent (PMD) was evaluated against Culicoides impunctatus in Scotland in comparison with deet. In human landing catches, both repellents still afforded 98% protection from biting 8 h after application of 0.5 ml to the forearm. A second trial looking at protection between 8 and 10 h after repellent application showed 99.5% protection for PMD and 97% for deet as compared with controls.