Merck
  • Evidence of chemical stimulation of hepatic metabolism by an experimental acetanilide (FOE 5043) indirectly mediating reductions in circulating thyroid hormone levels in the male rat.
Evidence of chemical stimulation of hepatic metabolism by an experimental acetanilide (FOE 5043) indirectly mediating reductions in circulating thyroid hormone levels in the male rat.
Fundamental and applied toxicology : official journal of the Society of Toxicology (1996-02-01)
W R Christenson, B D Becker, B S Wahle, K D Moore, P D Dass, S G Lake, D L Van Goethem, B P Stuart, G K Sangha, J H Thyssen
ABSTRACT
N-(4-Fluorophenyl)-N-(1-methylethyl)-2-[[5-(trifluoromethyl)-1,3, 4-thiadiazol-2-yl]oxy]acetamide (FOE 5043) is a new acetanilide-type herbicide undergoing regulatory testing. Previous work in this laboratory suggested that FOE 5043-induced reductions in serum thyroxine (T4) levels were mediated via an extrathyroidal site of action. The possibility that the alterations in circulating T4 levels were due to chemical induction of hepatic thyroid hormone metabolism was investigated. Treatment with FOE 5043 at a rate of 1000 ppm as a dietary admixture was found to significantly increase the clearance of [125I]T4 from the serum, suggesting an enhanced excretion of the hormone. In the liver, the activity of hepatic uridine glucuronosyl transferase, a major pathway of thyroid hormone biotransformation in the rat, increased in a statistically significant and dose-dependent manner; conversely, hepatic 5'-monodeiodinase activity trended downward with dose. Bile flow as well as the hepatic uptake and biliary excretion of [125I]T4 were increased following exposure to FOE 5043. Thyroidal function, as measured by the discharge of iodide ion in response to perchlorate, and pituitary function, as measured by the capacity of the pituitary to secrete thyrotropin in response to an exogenous challenge by hypothalamic thyrotropin releasing hormone, were both unchanged from the controlled response. These data suggest that the functional status of the thyroid and pituitary glands has not been altered by treatment with FOE 5043 and that reductions in circulating levels of T4 are being mediated indirectly through an increase in the biotransformation and excretion of thyroid hormone in the liver.