- Synthesis of ethyl arachidonate-19,19,20,20-d4 and ethyl dihomo-gamma-linolenate-19,19,20,20-d4.
Synthesis of ethyl arachidonate-19,19,20,20-d4 and ethyl dihomo-gamma-linolenate-19,19,20,20-d4.
Lipids (1993-09-01)
D L Luthria, H Sprecher
PMID8231662
ABSTRACT
Ethyl 5,8,11,14-eicosatetraenoate-19,19,20,20-d4 and ethyl 8,11,14-eicosatrienoate-19,19,20,20-d4 were synthesized by Grignard coupling of the methanesulfonyl ester of 2,5-undecadiyn-1-ol-10,10,11,11-d4 with 5,8-nonadiynoic acid and 8-nonynoic acid, respectively. The coupled products upon Lindlar reduction, followed by the preparation of their ethyl esters, yielded deuteriated ethyl arachidonate and ethyl dihomo-gamma-linolenate, which were completely characterized by 13C and 1H nuclear magnetic resonance and mass spectral analysis.