- [Moroxidine, an antiviral used for the treatment of zona (herpes zoster)].
[Moroxidine, an antiviral used for the treatment of zona (herpes zoster)].
Revue roumaine de virologie (Bucharest, Romania : 1990) (1993-01-01)
P Athanasiu, A Petrescu, V Vulcan
PMID8043474
ABSTRACT
Treatment with moroxidine, Romanian preparation with virustatic effects, was applied in 350 patients with different localisation herpes zoster lesions. Treatment had good effects, especially when it was applied early.