Merck
  • Dibutyltin oxide--phenyl isocyanate system for regioselective phenylcarbamoylation of the hydroxy-groups of ribonucleosides.
Dibutyltin oxide--phenyl isocyanate system for regioselective phenylcarbamoylation of the hydroxy-groups of ribonucleosides.
Nucleic acids symposium series (1980-01-01)
Y Ishido, I Hirao, K Itoh, K Tamaki, Y Araki
ABSTRACT
For partial phenylcarbamoylation of the hydroxy-groups of ribonucleosides, dibutyltin oxide--phenyl isocyanate system was found to be surperior to the bis(tributyltin) oxide--phenyl isocyanate system from the standpoint of reaction procedures including isolation of the products; the reaction was proved to occur with similar regioselectivity and to give the corresponding 5'-, 3'-, and 2'-O-phenylcarbamoyl derivatives in good yields, respectively, due to the conditions used.