Merck
  • Reduction of the scatter dose to the testicle outside the radiation treatment fields.
Reduction of the scatter dose to the testicle outside the radiation treatment fields.
International journal of radiation oncology, biology, physics (1982-10-01)
H Kubo, W U Shipley
ABSTRACT
A technique is described to reduce the dose to the contralateral testicle of patients with testis tumors during retroperitoneal therapy with 10 MV X rays. When a conventional clam-shell shielding device was used, the dose to the testis from the photons scattered by the patient and the collimator jaws was found to be about 1.6% of the prescribed midplane dose. A more substantial gonadal shield made of low melting point Ostalloy, that reduced further the dose from internal scattered X rays, was therefore designed. A 10 cm thick lead scrotal block above the scrotum immediately outside the field is shown to reduce the external scattered radiation to negligible levels. Using the shield and the block, it is possible to reduce the dose to the testicle to one-tenth of one percent of the prescribed midplane dose.