Merck
Fluoroimmunoassay of progesterone in human serum or plasma.
Clinical chemistry (1981-07-01)
B L Allman, F Short, V H James
ABSTRACT
We describe a fluoroimmunoassay for progesterone in serum or plasma. The assay involves use of an antiserum to progesterone-11-hemisuccinate and a labeled antigen prepared by linking fluoresceinamine to progesterone-3-carboxymethyloxime by use of a mixed-anhydride procedure. Serum or plasma samples are extracted with hexane. After incubation of a portion of the evaporated extract with antiserum and labeled antigen, antibody-bound and free antigen are separated and the fluorescence of the bound fraction is measured. Separation is effected either by using ammonium sulfate to precipitate liquid-phase antiserum or by using a solid-phase antiserum: antiserum covalently linked to magnetizable cellulose particles, which are separated with a magnet. With either separation technique, analytical recovery, linearity, and precision were acceptable and results compared satisfactorily with those obtained with a conventional radioimmunoassay. The solid-phase assay is more precise and more technically convenient, but the performance of both fluoroimmunoassays was adequate for clinical use in the detection of ovulation.