Merck
A new fluorometric assay method for quinolinic acid.
Analytical biochemistry (1983-05-01)
H Taguchi, S Koyama, Y Shimabayashi, K Iwai
ABSTRACT
A new fluorometric assay method for quinolinic acid is introduced in this study. Quinolinic acid-hydrazine complex, a stable fluorescent compound, is formed after heating quinolinic acid with hydrazine at 215-220 degrees C for 2 min. Fluorescence excitation and emission maxima of the complex are at 285 and 380 nm, respectively. This assay method is rapid and rather sensitive. It takes about 30 min to ascertain the amount of quinolinic acid as low as 50 ng. Specificity of this method is high among biological compounds. An ultrasensitive assay method for quinolinic acid (as low as 20 pg) with diphenylhydrazine instead of hydrazine is also found. After separating the quinolinic acid-diphenylhydrazine complex from residual diphenylhydrazine, this ultrasensitive assay method may be practically applicable.