Merck
  • Biological activity of 4-hydroxyisophthalic acid derivatives. II. Anilides with antimicrobial activity.
Biological activity of 4-hydroxyisophthalic acid derivatives. II. Anilides with antimicrobial activity.
Bollettino della Societa italiana di biologia sperimentale (1984-12-30)
E Piscopo, M V Diurno, M T Cataldi, G Scala, F Aliberti
ABSTRACT
A series of 1,3 -bis-anilides of 4-hydroxyisophthalic acid was prepared and tested for antibacterial and antifungal activity. The prepared compounds (I-XVIII), of general structure (A), (Formula: see text) where Xn = H (I); 2-F (II); 3-F (III); 4-F (IV); 2-Cl (V); 3-Cl (VI); 4-Cl (VII); 2-Br (VIII); 3-Br (IX); 4-Br (X); 2-J (XI); 3-J (XII); 4-J (XIII); 2,5-Cl2 (XIV); 2,4-Br2 (XV); 2,3,4-Cl3 (XVI), 2,4,5-Cl3 (XVII); 2,4,6-Cl3 (XVIII), were investigated for the purpose of determining the effect of halogen-substitution on the aniline rings of (A). All of these compounds were prepared in satisfactory hield by reaction of 4-hydroxyisophthalic acid with the appropriate aromatic amine at 175 degrees for 3 hours. The 1,3-bis-anilides prepared in this investigation were screened for antimicrobial activity by a disk-diffusion assay (Kirby-Bauer modified). The organisms used were laboratory cultures of S. aureus, B. subtilis, B. anthracis, M. paratuberculosis 607, E. coli Bb, S. typhi, S. typhimurium, S. paratyphi B, Pr. vulgaris, Kl. pneumoniae, Ps. aeruginosa, C. albicans, and A. niger. The results of this investigation indicated that most of the 1,3-bis-(halogen-anilides) of 4-hydroxyisophthalic acid had little or no antifungal activity "in vitro", while showed significant activity against Gram+ and Gram- bacteria. Some fluoro-derivatives showed inhibitory activity especially toward S. aureus and M. paratuberculosis. Iodo-derivatives showed broad-spectrum "in vitro" antimicrobial activity, and had some antifungal activity.