Merck
  • A new spin label method for the measurement of erythrocyte internal microviscosity.
A new spin label method for the measurement of erythrocyte internal microviscosity.
Biochimica et biophysica acta (1983-08-17)
D Daveloose, G Fabre, F Berleur, G Testylier, F Leterrier
ABSTRACT
A new spin-label method for the measurement of the internal microviscosity of erythrocyte is presented. The spin label used is 2,2',5,5'-tetramethyl-3-maleimidopyrrolidinyl-N-oxyl (MAL-5) which penetrates inside the red blood cell and binds covalently on cytoplasmic glutathione. After washing off the external label, 98% of the electron paramagnetic signal is due to the labelled glutathione. This signal allows one to measure the rotational correlation time of the label. A calibration curve established with spin-labelled glutathione in sucrose solutions of increasing viscosity is used to convert the measured rotation times into viscosity units. This method avoids the use of unphysiological salts like potassium ferricyanide, and permits the study of red blood cells in various suspension media. In normal human subjects, the mean value of microviscosity is 4.45 +/- 0.16 mPa . s at 20 degrees C in isotonic saline (25 subjects) and 6 +/- 0.25 mPa . s in plasma. The variations of microviscosity as a function of the osmolarity of the medium are explained according to a theoretical model taking into account the variations of the red blood cell volume and the viscometric properties of haemoglobin.