Merck
  • Cross-linking studies of adrenocortical cytochrome P-450scc. Evidence for a covalent complex with adrenodoxin and localization of the adrenodoxin-binding domain.
Cross-linking studies of adrenocortical cytochrome P-450scc. Evidence for a covalent complex with adrenodoxin and localization of the adrenodoxin-binding domain.
Biochimica et biophysica acta (1985-04-29)
V L Chashchin, I V Turko, A A Akhrem, S A Usanov
ABSTRACT
A cleavable cross-linking reagent, dimethyl-3,3'-dithiobispropionimidate, was used to study the molecular organization of adrenocortical cytochrome P-450scc. Extensive cross-linking was found to occur, resulting in the formation of heterologous oligomers up to octamer. The covalently cross-linked complex of adrenocortical cytochrome P-450scc with adrenodoxin has been obtained by using dimethyl-3,3'-dithiobispropionimidate. In the presence of NADPH and adrenodoxin reductase, electron transfer to cytochrome P-450scc occurs in the complex, and, in the presence of cholesterol, the latter effectively oxidizes to pregnenolone. By using covalently immobilized adrenodoxin and heterobifunctional reagent, N-succinimidyl-3-(2-pyridyldithio)propionate, the adrenodoxin-binding site was shown to be located in the heme-containing, catalytic domain of cytochrome P-450scc. The data obtained indicate the existence of two different sites on the adrenodoxin molecule that are responsible for the interaction with adrenodoxin reductase and cytochrome P-450scc. This is consistent with the model mechanism of electron transfer in the organized complex.