- [Theoretical conformation analysis of beta-casomorphin-5 molecule].
[Theoretical conformation analysis of beta-casomorphin-5 molecule].
Molekuliarnaia biologiia (1986-11-01)
N A Akhmedov
PMID3807911
ABSTRACT
The spatial structure of beta-casomorphin-5 molecule--H-Tyr-Pro-Phe-Pro-Gly-OH has been investigated by the theoretical conformational analysis method. The results indicate that the conformational properties of the molecule can be represented by a large number of structures having twelve different backbone forms.