Merck
  • Long-term memory formation in chicks is blocked by 2-deoxygalactose, a fucose analog.
Long-term memory formation in chicks is blocked by 2-deoxygalactose, a fucose analog.
Behavioral and neural biology (1987-09-01)
S P Rose, R Jork
ABSTRACT
When day-old chicks are trained on a passive avoidance task there is enhanced synthesis of glycoproteins. Bilateral intracerebral injections of 20 mumole of 2-deoxygalactose (2-D-gal), administered just before and just after training on the task, produce amnesia for the avoidance. Amnesia develops slowly over the first hour and persists for at least 24 h subsequently. If 2-D-gal injections are administered 4 h prior to the training or delayed for 3 h after training, no amnesia occurs. Apart from a brief initial suppression of pecking following injection there are no effects of 2-D-gal on other observed behaviors of the birds. Within the first hour this dose of 2-D-gal inhibits [3H]fucose incorporation into acid-insoluble material by 26% (or 68%, calculated relative to free pool fucose). The amnestic effect of 2-D-gal is not shown by galactose, glucose, fucose, or 2-D-glucose. Injecting 40 mumole of galactose simultaneously with the 2-D-gal abolishes the 2-D-gal-induced amnesia; 40 mumole of fucose, however, does not abolish the amnesia. The utility of 2-D-gal as an agent for analyzing the role of glycoproteins in memory formation is discussed.