Merck
  • Effect of human urinary trypsin inhibitor on granulocyte elastase activity.
Effect of human urinary trypsin inhibitor on granulocyte elastase activity.
Research communications in chemical pathology and pharmacology (1987-02-01)
M Ogawa, S Nishibe, T Mori, S Neumann
ABSTRACT
A human urinary trypsin inhibitor preparation, MR-20, strongly inhibited granulocyte elastase activity. The concentration of MR-20 causing 50% inhibition of the hydrolysis of Suc(Ala) 3pNA by granulocyte elastase was estimated to be 6.4 X 10(-8) M (11 U/ml).