Merck
  • Aminothiadiazole (NSC 4728) in patients with advanced nonsquamous carcinoma of the cervix. A phase II study of the Gynecologic Oncology Group.
Aminothiadiazole (NSC 4728) in patients with advanced nonsquamous carcinoma of the cervix. A phase II study of the Gynecologic Oncology Group.
American journal of clinical oncology (1989-10-01)
R F Asbury, J A Blessing, P J DiSaia, J Malfetano
ABSTRACT
Twenty-six evaluable patients with advanced nonsquamous cell cervical cancer were treated with aminothiadiazole at a dosage of 125 mg/m2 weekly. Sixteen had received prior chemotherapy. Two patients had complete responses neither had received prior chemotherapy; nine had stable disease; 15 had increasing disease. There was a single life-threatening toxic episode. Aminothiadiazole used in this dosage and schedule has minimal activity in previously treated patients with nonsquamous cell carcinoma.