Merck
  • Chemotherapy of caecal coccidiosis: efficacy of toltrazuril, sulphaquinoxaline/pyrimethamine and amprolium/ethopabate, given in drinking water, against field isolates of Eimeria tenella.
Chemotherapy of caecal coccidiosis: efficacy of toltrazuril, sulphaquinoxaline/pyrimethamine and amprolium/ethopabate, given in drinking water, against field isolates of Eimeria tenella.
Research in veterinary science (1989-05-01)
H D Chapman
ABSTRACT
Treatment with toltrazuril, sulphaquinoxaline/pyrimethamine and amprolium/ethopabate prevented mortality in chickens infected with field isolates of Eimeria tenella. Amprolium/ethopabate was the most effective drug in reducing lesions caused by the parasites. Few oocysts of E tenella were produced in birds medicated with sulphaquinoxaline/pyrimethamine or amprolium/ethopabate and none in those medicated with toltrazuril.