Merck
  • Facile synthesis of monoacetylated spermidines, illustrating selective deacetylation and application of a common precursor.
Facile synthesis of monoacetylated spermidines, illustrating selective deacetylation and application of a common precursor.
Acta chemica Scandinavica (Copenhagen, Denmark : 1989) (1989-11-01)
M Lurdes, S Almeida, L Grehn, U Ragnarsson
ABSTRACT
The synthesis of all three monoacetylated spermidines is reported. N4-Acetylspermidine was obtained in four steps from spermidine via the triacetylated intermediate by selective deacetylation after exhaustive t-butoxycarbonylation as well as directly from a previously described protected precursor. N1-Acetylspermidine and N8-acetylspermidine were both obtained in four simple protection/deprotection steps from a common, selectively protected compound, thus illustrating the versatility of the latter.