Merck
  • Rhodium-catalyzed conversion of furans to highly functionalized pyrroles.
Rhodium-catalyzed conversion of furans to highly functionalized pyrroles.
Journal of the American Chemical Society (2013-03-13)
Brendan T Parr, Samantha A Green, Huw M L Davies
ABSTRACT
The synthesis of highly functionalized pyrroles has been achieved by reaction of rhodium-stabilized imino-carbenes with furans. The reaction features an initial [3+2] annulation to form bicyclic hemiaminals, followed by ring opening to generate trisubstituted pyrroles.