- A randomized controlled trial of 1% aqueous chlorhexidine gluconate compared with 10% povidone-iodine for topical antiseptic in neonates: effects on blood culture contamination rates.
A randomized controlled trial of 1% aqueous chlorhexidine gluconate compared with 10% povidone-iodine for topical antiseptic in neonates: effects on blood culture contamination rates.
Infection control and hospital epidemiology (2013-03-08)
Pracha Nuntnarumit, Nartsiri Sangsuksawang
PMID23466918
ABSTRACT
We conducted a randomized controlled trial in neonates with birth weight greater than or equal to 1,500 g that compared 1% aqueous chlorhexidine gluconate (CHG) with 10% povidone-iodine (PI) as a topical antiseptic. We found 1% CHG to be more effective than 1% PI in reducing blood culture contamination rates, and no contact dermatitis was observed.